4-(Trifluoromethyl)-1-indanone 97%

Supplier: Ambeed
Warning

A151593-250MG A151593-1G A151593-5G A151593-10G
103804-278EA 16 USD
103804-278 103842-430 76973-136 77318-006 77318-008 77881-173
4-(Trifluoromethyl)-1-indanone 97%
4-(Trifluoromethyl)-1-indanone

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O
Melting Pt: 38…42 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD07772121
CAS Number: 68755-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR