(4-(Trifluoromethoxy)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethoxy)phenylacetic acid

A103678-10G A103678-25G A103678-5G
103796-596EA 7 USD
103796-596 103822-304 77283-142 77283-144 77309-172
(4-(Trifluoromethoxy)phenyl)acetic acid 97%
(4-(Trifluoromethoxy)phenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(OCF₃))CH₂COOH
MW: 220.15 g/mol
Boiling Pt: 260.6 °C (760 mm Hg)
Melting Pt: 85…88 °C
Flash Pt: 111.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00061239
CAS Number: 4315-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR