4-(Trifluoromethoxy)cinnamic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethoxy)cinnamic Acid

A770069-1G A770069-25G A770069-5G A770069-10G
103840-930EA 52 USD
103840-930 76845-170 76845-172 76845-174 76880-102
4-(Trifluoromethoxy)cinnamic acid 97%
4-(Trifluoromethoxy)cinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₃
MW: 232.1600 g/mol
Melting Pt: 178 °C
Storage Temperature: Ambient
MDL Number: MFCD00066338
CAS Number: 783-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR