4-(Trifluoromethoxy)benzonitrile 98%

Supplier: Ambeed
Warning

A647388-25G A647388-5G A647388-100G A647388-10G
103840-382EA 9 USD
103840-382 76844-544 76844-546 76980-784 77882-486
4-(Trifluoromethoxy)benzonitrile 98%
4-(Trifluoromethoxy)benzonitrile

Precaución: For research use only

Formula: C₈H₄F₃NO
Boiling Pt: 187.6 °C (760 mm Hg)
Flash Pt: 67.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00039474
CAS Number: 332-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR