4-(Trifluoromethoxy)benzoic acid 95%

Supplier: Ambeed
Warning

A185621-100G A185621-10G A185621-500G A185621-1G A185621-25G A185621-5G
103815-866EA 5 USD
103815-866 103846-436 76769-722 76769-724 76769-726 77301-984 77301-986 77301-988
4-(Trifluoromethoxy)benzoic acid 95%
4-(Trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 230.2 °C (760 mm Hg)
Melting Pt: 150…154 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002541
CAS Number: 330-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR