4-(Trifluoromethoxy)-1,2-phenylenediamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Trifluoromethoxy)benzene-1,2-diamine
A746782-1G
A746782-100MG
A746782-250MG
103840-836EA
89
USD
103840-836
76845-066
77146-204
77146-206
4-(Trifluoromethoxy)-1,2-phenylenediamine 98%
4-(Trifluoromethoxy)-1,2-phenylenediamine
Precaución: for Research Use Only
Formula:
C₇H₇F₃N₂O MW: 192.14 g/mol Flash Pt: 104.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094278 CAS Number: 658-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...