4'-(Trifluoromethoxy)-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A347049-100MG A347049-250MG
103825-950EA 164 USD
103825-950 77115-080 77115-082
4'-(Trifluoromethoxy)-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid 95%
4'-(Trifluoromethoxy)-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₈F₆O₃
MW: 350.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18322456
CAS Number: 1261503-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR