4-(Tetrahydropyran-4-yl)phenylamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Oxan-4-yl)aniline

A114881-250MG A114881-5G
103798-492EA 80 USD
103798-492 103824-334 76971-110 77311-152
4-(Tetrahydropyran-4-yl)phenylamine 95%
4-(Tetrahydropyran-4-yl)phenylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.2429 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11506022
CAS Number: 62071-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR