4-tert-Butylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(tert-Butyl)pyridine
A234255-100G
A234255-10G
A234255-500G
A234255-1G
A234255-25G
A234255-5G
103810-934EA
5
USD
103810-934
103850-692
76771-216
76771-218
76771-220
77089-670
77089-672
77089-674
4-tert-Butylpyridine 98%
4-tert-Butylpyridine
Precaución: for Research Use Only
Formula:
C₉H₁₃N MW: 135.21 Boiling Pt: 196 Melting Pt: -0.9 Density: 0.9219 Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006435 CAS Number: 3978-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...