4-tert-Butylbenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A155429-100G
A155429-10G
A155429-500G
A155429-25G
A155429-5G
103842-958EA
5
USD
103842-958
76973-288
76973-290
76973-292
77318-676
77318-678
4-tert-Butylbenzonitrile 98%
4-tert-Butylbenzonitrile
Precaución: for Research Use Only
Formula:
C₁₁H₁₃N Boiling Pt: 245.6 °C (760 mm Hg) Flash Pt: 102.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075840 CAS Number: 4210-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...