4-tert-Butyl-o-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butyl)benzene-1,2-diamine, 4-tert-Butylbenzene-1,2-diamine, 4-tert-Butyl-1,2-phenylenediamine, 1,2-Diamino-4-(dimethylethyl)benzene, 1,2-Diamino-4-tert-butylbenzene

A133730-10G A133730-250MG A133730-25G A133730-1G
103801-908EA 16 USD
103801-908 76688-730 76688-732 77298-654 77314-592
4-tert-Butyl-o-phenylenediamine 97%
4-tert-Butyl-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
Boiling Pt: 290.5 °C (760 mm Hg)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00052695
CAS Number: 68176-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR