4-tert-Butylaniline 98%
Supplier: Ambeed
|
Warning
|
A187069-100G
A187069-10G
A187069-500G
A187069-25G
103809-210EA
105
USD
103809-210
103846-578
76690-716
76690-718
76690-720
77302-060
4-tert-Butylaniline 98%
4-tert-Butylaniline
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N Boiling Pt: 90…93 °C (3 mm Hg) Melting Pt: 15…16 °C Flash Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007899 CAS Number: 769-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...