4-(tert-Butyl)-2-ethoxybenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A264121-1G
A264121-25G
A264121-5G
A264121-100G
A264121-250MG
103812-654EA
10
USD
103812-654
103852-546
77092-200
77092-202
77092-204
77289-290
77289-292
4-(tert-Butyl)-2-ethoxybenzoic acid 97%
4-(tert-Butyl)-2-ethoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₈O₃ MW: 222.280 g/mol Boiling Pt: 329.6 °C (760 mm Hg) Flash Pt: 118.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD07366640 CAS Number: 796875-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...