4-tert-Butyl-2,6-dimethylaniline 98%

Supplier: Ambeed
Danger

Synonyms: 4-(tert-Butyl)-2,6-dimethylaniline

A114084-250MG A114084-1G A114084-5G A114084-25G A114084-10G
103798-344EA 23 USD
103798-344 76718-240 77311-010 77570-770 77592-212 77878-854
4-tert-Butyl-2,6-dimethylaniline 98%
4-tert-Butyl-2,6-dimethylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉N
Boiling Pt: 263.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00052181
CAS Number: 42014-60-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR