Thiomorpholine-3-carboxylic acid, N-BOC protected 95%
Supplier: Ambeed
|
Warning
|
A174109-250MG
A174109-1G
A174109-5G
103806-918EA
92
USD
103806-918
103845-216
76690-262
77082-504
77301-316
Thiomorpholine-3-carboxylic acid, N-BOC protected 95%
Thiomorpholine-3-carboxylic acid, N-BOC protected
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄S Boiling Pt: 398.6 °C (760 mm Hg) Melting Pt: 145…146 °C Flash Pt: 194.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02683067 CAS Number: 128453-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...