4-(tert-Butoxycarbonyl)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: mono-(tert-Butyl) terephthalate

A206579-100MG A206579-250MG A206579-10G
103817-716EA 15 USD
103817-716 103848-392 76721-578 76721-580 77550-322
4-(tert-Butoxycarbonyl)benzoic acid 95%
4-(tert-Butoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
Boiling Pt: 351.8 °C (760 mm Hg)
Melting Pt: 209…212 °C
Storage Temperature: Ambient
MDL Number: MFCD04037901
CAS Number: 20576-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR