4-(tert-Butoxy)benzeneboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butoxy)phenylboronic Acid, 4-tert-Butoxyphenylboronic acid

A224868-100G A224868-250MG A224868-25G A224868-5G
103819-138EA 12 USD
103819-138 103850-030 77000-020 77000-022 77000-024 77088-696
4-(tert-Butoxy)benzeneboronic acid 95%
4-(tert-Butoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Boiling Pt: 328.1±44.0 °C (760 mm Hg)
Flash Pt: 152.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094965
CAS Number: 176672-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR