tert-Butyl hydrogen succinate 97%

Supplier: Ambeed
Warning

A327918-100G A327918-10G A327918-500G A327918-1G A327918-25G A327918-5G
103825-480EA 5 USD
103825-480 103836-430 76724-076 76724-078 76724-080 76839-398 76839-400 76839-402
tert-Butyl hydrogen succinate 97%
tert-Butyl hydrogen succinate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
Boiling Pt: 272.4 °C (760 mm Hg)
Melting Pt: 51…54 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273441
CAS Number: 15026-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR