4-(Pyridin-4-yl)aniline 97%

Supplier: Ambeed
Warning

A167172-250MG A167172-1G A167172-5G A167172-100MG
103806-142EA 37 USD
103806-142 103844-404 77286-642 77300-878 77300-880 77748-108
4-(Pyridin-4-yl)aniline 97%
4-(Pyridin-4-yl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
MW: 170.2105 g/mol
Melting Pt: 232…234 °C
Flash Pt: 172.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00970053
CAS Number: 13296-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR