4-(Pyridin-3-yloxy)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-3-yloxy)-phenylamine

A181990-250MG A181990-1G
103807-676EA 56 USD
103807-676 103846-046 76784-620 77083-576
4-(Pyridin-3-yloxy)aniline 95%
4-(Pyridin-3-yloxy)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.2100 g/mol
Boiling Pt: 346.2 °C (760 mm Hg)
Flash Pt: 163.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07186369
CAS Number: 80650-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR