4-(Pyridin-3-yl)pyrimidin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Pyridinyl)-2-pyrimidinamine

A119148-10G A119148-25G
103799-172EA 26 USD
103799-172 77284-722 77284-724
4-(Pyridin-3-yl)pyrimidin-2-amine 97%
4-(Pyridin-3-yl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₉H₈N₄
MW: 172.19 g/mol
Boiling Pt: 439.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01317832
CAS Number: 66521-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR