4-(Pyridin-2-yl)phenol 95%

Supplier: Ambeed
Warning

A124023-10G A124023-25G A124023-1G A124023-5G
103800-474EA 10 USD
103800-474 76870-394 76870-396 77297-966 77297-968
4-(Pyridin-2-yl)phenol 95%
4-(Pyridin-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464189
CAS Number: 51035-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR