4-(4-Piperidyl)pyridine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-(4-Pyridyl)piperidine, 4-(Piperidin-4-yl)pyridine

A134091-1G A134091-250MG
103801-974EA 156 USD
103801-974 77314-692 77683-191
4-(4-Piperidyl)pyridine 97%, Ambeed.Inc
4-(4-Piperidyl)pyridine

Precaución: For research use only.

Formula: C₁₀H₁₄N₂
MW: 162.24 g/mol
Boiling Pt: 292 °C (760 mm Hg)
Melting Pt: 86…88 °C
Flash Pt: 120.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05863877
CAS Number: 581-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR