4-(4-Cyanophenyl)piperidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-piperidin-4-ylbenzonitrile, 4-(Piperidin-4-yl)benzonitrile

A171684-1G A171684-5G A171684-250MG
103806-656EA 133 USD
103806-656 103844-932 77082-102 77082-104 77286-896
4-(4-Cyanophenyl)piperidine 95%
4-(4-Cyanophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂
MW: 186.253 g/mol
Boiling Pt: 333.6 °C (760 mm Hg)
Flash Pt: 155.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03426383
CAS Number: 149554-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR