4-(Octylamino)pyridine 98%

Supplier: Ambeed
Warning

A210356-250MG A210356-1G A210356-5G
103817-996EA 13 USD
103817-996 103848-690 76814-512 77087-060 77087-062
4-(Octylamino)pyridine 98%
4-(Octylamino)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂
MW: 206.3300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743883
CAS Number: 64690-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR