4-(N-Boc-aminomethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-(N-Boc-Aminomethyl)aniline

A234590-100MG A234590-10G A234590-250MG A234590-1G A234590-25G A234590-5G
103810-962EA 12 USD
103810-962 103850-718 76771-224 76771-226 76771-228 77089-710 77089-712 77303-986
4-(N-Boc-aminomethyl)aniline 98%
4-(N-Boc-aminomethyl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.288 g/mol
Melting Pt: 75…78 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD03001716
CAS Number: 94838-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR