4-(N,N-Diphenylamino)benzaldehyde 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Diphenylaminobenzaldehyde
A331814-100G
A331814-10G
A331814-1G
A331814-25G
A331814-5G
103825-588EA
7
USD
103825-588
103836-554
77032-880
77032-882
77306-420
77306-422
77306-424
4-(N,N-Diphenylamino)benzaldehyde 96%
4-(N,N-Diphenylamino)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₉H₁₅NO MW: 273.33 g/mol Boiling Pt: 436.8 °C (760 mm Hg) Melting Pt: 129…133 °C Flash Pt: 171.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145131 CAS Number: 4181-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...