4-(m-Tolylamino)pyridine-3-sulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: 4-(m-Tolylamino)-3-sulphamoylpyridine, 4-(3-Methylphenyl)aminopyridine-3-sulphonamide, Torsemide Related Compound A, 4-(3-Methylphenyl)amino-3-sulphamoylpyridine

A123622-100G A123622-1G A123622-250MG A123622-25G
76766-750EA 698.08 USD
76766-750 76766-752 76766-754 76766-756
4-(m-Tolylamino)pyridine-3-sulfonamide 95%
4-(m-Tolylamino)pyridine-3-sulfonamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₂S
MW: 263.32 g/mol
Boiling Pt: 475.8 °C (760 mm Hg)
Melting Pt: 162…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00661332
CAS Number: 72811-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR