4-(Morpholinomethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Morpholinomethyl)benzeneboronic acid, [4-(Morpholin-4-ylmethyl)phenyl]boronic acid

A175603-10G A175603-1G A175603-5G A175603-100MG
103807-066EA 39 USD
103807-066 103845-378 76974-238 76974-240 77082-720 77082-722 77671-496
4-(Morpholinomethyl)phenylboronic acid 98%
4-(Morpholinomethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
Boiling Pt: 382.3 °C (760 mm Hg)
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD06801707
CAS Number: 279262-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR