4-Morpholin-4-ylmethylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Morpholinomethyl)benzoic acid

A148042-10G A148042-1G
103803-870EA 16 USD
103803-870 103841-986 77286-024 77317-422
4-Morpholin-4-ylmethylbenzoic acid 98%
4-Morpholin-4-ylmethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD01167722
CAS Number: 62642-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR