4-(Methylthio)phenylacetic acid 97%

Supplier: Ambeed
Warning

A344996-1G A344996-5G A344996-100G A344996-10G
103825-914EA 5 USD
103825-914 103836-896 76839-948 76839-950 77251-454 77251-456
4-(Methylthio)phenylacetic acid 97%
4-(Methylthio)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
MW: 182.24 g/mol
Boiling Pt: 337.4±25.0 °C (760 mm Hg)
Melting Pt: 97…98 °C
Flash Pt: 157.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192325
CAS Number: 16188-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR