4-(Methylthio)benzaldehyde 98%

Supplier: Ambeed
Warning

A104523-100G A104523-10G A104523-1G A104523-25G A104523-5G
103822-446EA 6 USD
103822-446 77283-232 77283-234 77283-236 77283-238 77283-240
4-(Methylthio)benzaldehyde 98%
4-(Methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈OS
MW: 152.22 g/mol
Boiling Pt: 86…90 °C (1 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006948
CAS Number: 3446-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR