4-(Methylsulfonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

A183090-1G A183090-5G A183090-100G A183090-10G A183090-25G
103815-604EA 6 USD
103815-604 103846-168 77083-754 77083-756 77191-752 77191-754 77301-822
4-(Methylsulfonyl)phenylboronic acid 98%
4-(Methylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₄S
MW: 200.02 g/mol
Boiling Pt: 445.5 °C (760 mm Hg)
Melting Pt: 275…277 °C
Flash Pt: 223.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01630820
CAS Number: 149104-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR