4'-(Methylsulfonyl)acetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 4'-(Methylsulphonyl)acetophenone

A299585-25G A299585-5G A299585-100G A299585-10G A299585-500G A299585-1000G
103835-382EA 5 USD
103835-382 76838-074 76838-076 77250-772 77250-774 77250-776 77746-562
4'-(Methylsulfonyl)acetophenone 97%
4'-(Methylsulphonyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₃S
MW: 198.24 g/mol
Boiling Pt: 385.4 °C (760 mm Hg)
Melting Pt: 126…129 °C
Flash Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD00025072
CAS Number: 10297-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR