4-(Methylamino)butanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A102920-10G A102920-25G A102920-5G A102920-100G
103822-150EA 10 USD
103822-150 77283-012 77283-014 77283-016 77747-766
4-(Methylamino)butanoic acid hydrochloride 97%
4-(Methylamino)butanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₂
MW: 153.6 g/mol
Boiling Pt: 269.6 °C (760 mm Hg)
Flash Pt: 116.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00012612
CAS Number: 6976-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR