4-(Methylamino)butan-1-ol 97%

Supplier: Ambeed
Warning

A113629-100MG A113629-1G A113629-5G A113629-25G
103798-260EA 52 USD
103798-260 103824-100 77284-306 77310-912 77310-914 77747-826
4-(Methylamino)butan-1-ol 97%
4-(Methylamino)butan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 167.18 °C (760 mm Hg)
Flash Pt: 67.237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00673140
CAS Number: 42042-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR