4-(Hydroxymethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxybenzylalcohol, HMBA Linker, HMBA

A165011-100G A165011-10G A165011-1G A165011-25G A165011-5G A165011-500G
103805-932EA 9 USD
103805-932 103844-190 77247-764 77247-766 77300-750 77300-752 77300-754 77615-358
4-(Hydroxymethyl)benzoic acid 98%
4-(Hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 347.4 °C (760 mm Hg)
Melting Pt: 182…185 °C
Flash Pt: 178.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00017598
CAS Number: 3006-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR