4-(Hydroxymethyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-(Hydroxymethyl)benzeneboronic acid

A114457-100G A114457-10G A114457-25G A114457-500G A114457-5G
103798-412EA 5 USD
103798-412 103824-250 77189-254 77189-256 77189-258 77189-260 77297-310
4-(Hydroxymethyl)phenylboronic acid 96%
4-(Hydroxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
Boiling Pt: 362 °C (760 mm Hg)
Melting Pt: 251…256 °C
Flash Pt: 172.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00792672
CAS Number: 59016-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR