(4-Dimethylaminophenyl)di-tert-butylphosphine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Di-tert-butylphosphino)-N,N-dimethylaniline

A186373-250MG A186373-1G A186373-25G A186373-5G A186373-100G
103815-934EA 8 USD
103815-934 103846-514 76985-908 77084-170 77084-172 77084-174 77683-476
(4-Dimethylaminophenyl)di-tert-butylphosphine 97%
(4-Dimethylaminophenyl)di-tert-butylphosphine

Precaución: For research use only.

Formula: C₁₆H₂₈NP
MW: 265.38 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD09265102
CAS Number: 932710-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR