4-(Dimethylamino)phenyldiphenylphosphine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Dimethylamino)triphenylphosphine, 4-(Diphenylphosphino)-N,N-dimethylaniline

A152681-100G A152681-10G A152681-1G A152681-25G A152681-5G
103804-420EA 9 USD
103804-420 103842-592 76871-668 76871-670 77318-220 77318-222 77318-224
4-(Dimethylamino)phenyldiphenylphosphine 97%
4-(Dimethylamino)phenyldiphenylphosphine

Precaución: for Research Use Only

Formula: C₂₀H₂₀NP
MW: 305.36 g/mol
Boiling Pt: 417.5 °C (760 mm Hg)
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00192068
CAS Number: 739-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR