4-(Dimethylamino)phenol 97%

Supplier: Ambeed
Warning

A225308-1G A225308-5G
103819-170EA 7 USD
103819-170 103850-064 77088-750 77088-752
4-(Dimethylamino)phenol 97%
4-(Dimethylamino)phenol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.179 g/mol
Boiling Pt: 260 °C (760 mm Hg)
Melting Pt: 138…141 °C
Flash Pt: 136 °C
Storage Temperature: Freezer
MDL Number: MFCD01707548
CAS Number: 619-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR