4-(Dimethylamino)cyclohexanone 95%
Supplier: Ambeed
|
Warning
|
A190293-1G
A190293-5G
A190293-250MG
A190293-25G
103816-282EA
15
USD
103816-282
103846-884
77084-672
77084-674
77164-258
77302-222
4-(Dimethylamino)cyclohexanone 95%
4-(Dimethylamino)cyclohexanone
Precaución: for Research Use Only
Formula:
C₈H₁₅NO MW: 141.2100 g/mol Boiling Pt: 213.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09037892 CAS Number: 40594-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...