4-(Dimethylamino)cyclohexanol 96%

Supplier: Ambeed
Warning

A193336-1G A193336-5G A193336-100MG A193336-250MG
103816-524EA 34 USD
103816-524 103847-138 77085-038 77085-040 77164-336 77164-338
4-(Dimethylamino)cyclohexanol 96%
4-(Dimethylamino)cyclohexanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO
Boiling Pt: 216.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09037891
CAS Number: 61168-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR