4-(Dimethylamino)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

A128847-100G A128847-1G A128847-500G A128847-25G
103808-390EA 56 USD
103808-390 77026-950 77026-952 77026-954 77549-534
4-(Dimethylamino)benzaldehyde 97%
4-(Dimethylamino)benzaldehyde

Precaución: for Research Use Only

Formula: (CH₃)₂NC₆H₄CHO
Boiling Pt: 266.5 °C (760 mm Hg)
Melting Pt: 72…75 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR