4-(Difluoromethoxy)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A191537-10G
A191537-250MG
A191537-1G
A191537-25G
103816-380EA
6
USD
103816-380
103846-984
77236-922
77236-924
77302-262
77302-264
4-(Difluoromethoxy)benzoic acid 98%
4-(Difluoromethoxy)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆F₂O₃ Boiling Pt: 272.1 °C (760 mm Hg) Melting Pt: 169…171 °C Flash Pt: 118.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00221454 CAS Number: 4837-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...