4-(Difluoromethoxy)benzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A189059-100G
A189059-1G
A189059-25G
A189059-5G
A189059-10G
103816-152EA
9
USD
103816-152
103846-746
76873-006
77084-496
77084-498
77084-500
77671-676
4-(Difluoromethoxy)benzaldehyde 97%
4-(Difluoromethoxy)benzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆F₂O₂ MW: 172.13 g/mol Boiling Pt: 234.7 °C (760 mm Hg) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042252 CAS Number: 73960-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...