4-(Difluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

A534222-25G A534222-5G A534222-1G A534222-10G
103839-704EA 14 USD
103839-704 76843-718 76843-720 77004-972 77594-648
4-(Difluoromethoxy)aniline 98%
4-(Difluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 224.1 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085005
CAS Number: 22236-10-8
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR