4-(Chlorosulfonyl)benzoic acid 95%
Supplier: Ambeed
|
Danger
|
A146432-10G
A146432-25G
103803-666EA
10
USD
103803-666
103841-756
77137-558
77299-518
4-(Chlorosulfonyl)benzoic acid 95%
4-(Chlorosulphonyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅ClO₄S Melting Pt: 233…235 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007448 CAS Number: 10130-89-9 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...