4-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride ≥95%

Supplier: Ambeed
Danger

A228507-100MG A228507-250MG
103819-400EA 98 USD
103819-400 103850-308 77890-341 77890-342
4-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride ≥95%
4-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₅H₈Cl₂N₂
MW: 167.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697625
CAS Number: 17289-30-4
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR