4-Picolyl bromide hydrobromide 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-(Bromomethyl)pyridine hydrobromide
A469649-1G
A469649-25G
A469649-5G
A469649-10G
103828-014EA
6
USD
103828-014
103839-088
76842-994
76842-996
76842-998
77291-854
77291-856
4-Picolyl bromide hydrobromide 95%
4-Picolyl bromide hydrobromide
Precaución: for Research Use Only
Formula:
C₆H₇Br₂N Melting Pt: 189…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863545 CAS Number: 73870-24-3 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...